Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BINA, Tocris Bioscience™

Catalog No. p-4816930 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective positive allosteric modulator of mGlu2

Selective positive allosteric modulator of mGlu2 (EC50 = 33.2 nM in CHO cells expressing human mGlu2). Displays no effect on glutamate-induced activation of other mGlu receptor subtypes. Exhibits antipsychotic and anxiolytic properties in mice.

Chemical Identifiers

CAS 866823-73-6
Molecular Formula C30H30O4
Molecular Weight (g/mol) 454.566
InChI Key KMKBEESNZAPKMP-UHFFFAOYSA-N
Synonym biphenylindanone a, bina, biphenyl-indanone a, 3'-2-cyclopentyl-2,3-dihydro-6,7-dimethyl-1-oxo-1h-inden-5-yl oxy methyl-1,1'-biphenyl-4-carboxylic acid, 3'-2-cyclopentyl-6,7-dimethyl-1-oxo-2,3-dihydro-1h-inden-5-yl oxy methyl-1,1'-biphenyl-4-carboxylic acid, d07mhl, mrlsd 230, 4-3-2-cyclopentyl-6,7-dimethyl-1-oxo-2,3-dihydroinden-5-yl oxymethyl phenyl benzoic acid, biphenyl-indanone a hplc , powder, 4-3-2-cyclopentyl-6,7-dimethyl-1-oxo-indan-5-yl oxymethyl phenyl benzoic acid
PubChem CID 9868580
IUPAC Name 4-[3-[(2-cyclopentyl-6,7-dimethyl-1-oxo-2,3-dihydroinden-5-yl)oxymethyl]phenyl]benzoic acid
SMILES CC1=C(C=C2CC(C(=O)C2=C1C)C3CCCC3)OCC4=CC=CC(=C4)C5=CC=C(C=C5)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 454.56
Percent Purity >98%
Chemical Name or Material BINA
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.