Learn More
Research Products International Corp Bio-Filtrators, PVDF, 250ml, 12/cs
SDP
Supplier:  Research Products International Corp 192425
Encompass
S Vacuum Bio-Filtrators have a built in 50mm diameter filter with a pore size of 0.22µm. Ideal for use in filtering biological fluids, culture media and other aqueous solutions. Item has a wide bottle opening for easy access and pouring the product. The funnel and bottle are manufactured from optically clear polystyrene. The filter adapter is manufactured from 100% virgin polypropylene. Graduations are engraved to provide more accuracy The cap for the receiver bottle is S and individually wrapped The reservoir has a large cap for easy pouring Hose connector fits multiple hose diameters All product come certified non-pyrogenic and Gamma irradiated Individually wrapped, peel-open packaging Units are printed on the case for easy traceabilityShipping Weight: 6 lbsShipping Dimensions: 17.00 x 14.50 x 10.00Product usually ships within 24 hours.Research or further manufacturing use only, not for food or drug use.
Catalog No. 50-222-9465
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.