Learn More
- Cost effective micro slide viewer
- High quality images
- Light weight
- Cordless
- Easy to use
Specifications
Grade
K to 5
Description
Cordless Bio Viewer
Format
Kit
Product Type
Slide Viewer
Color
Yellow
Subject
Biology - Life Science
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.