Learn More
Alkali Scientific BioCleanC™ Chemotherapy Protective Apron with Sleeves, Size Large, 40 /cs
Supplier: Alkali Scientific A933
Providing protection against a range of chemotherapy drugs, the BioClean-C™ Chemotherapy Protective Apron with Sleeves is manufactured from lightweight, low-linting CleanTough™ blue material, and features tie tapes at the rear and an adjustable neck fastening for easy donning and comfort. Tested against permeation standard ASTM F739-12.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.