Learn More
Alkali Scientific BioCleanC™ Chemotherapy Protective Apron with Sleeves, Size Medium, 40 /cs
SDP
Supplier:  Alkali Scientific A932
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Providing protection against a range of chemotherapy drugs, the BioClean-C™ Chemotherapy Protective Apron with Sleeves is manufactured from lightweight, low-linting CleanTough™ blue material, and features tie tapes at the rear and an adjustable neck fastening for easy donning and comfort. Tested against permeation standard ASTM F739-12.
Catalog No. 50-238-9257