Learn More
Aniara Diagnostica LLC BIOPHEN PLASMA CALIBRATOR RUO
Supplier:  Aniara Diagnostica LLC A222101RUO
Encompass_Preferred
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. Normal human citrated plasma, lyophilized. Assigned (assayed) values for PC, PS, AT, PT, aPTT, Fibrinogen, Prothrombin, FVII+X, Factors V, VII, VIII, IX, X, XI, XII & Plasminogen. Conc's established against NIBSC Std. 12x1 mL vials. RUO.
Catalog No. 50-100-8893
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.