Learn More
Thomson Instrument Company Bioreactor Exhaust Cap for 1.6L, Sterile | Use with p/n 931113 for Bioreactors | CS8
Supplier: Thomson Instrument Company 9317138
Thomson Instrument Company Biorecator Exhaust Cap for 1.6L Optimum Growth™ Flasks are born out of biopharmaceutical companies need to eliminate the risk of contamination and remain GMP compliant. The multiport flasks have a one-way port for sampling, and a two-way port for feeding and transfer. This allows for completely closed system aseptic processing while feeding, sampling, transferring, or inoculating. Thomson Optimum Growth™ Flasks are ideal for mammalian or insect cell culture, and available in 125mL, 250mL, 500mL, 1.6L, and 5L sizes. Shake flasks hold 2-3x more media compared to traditional shake flasks, greatly increasing shaker efficiency. These flasks feature a unique low shear baffle to increase aeration, while maintaining cell viability and improving yields. The Optimum Growth™ System consists of high efficiency standard and specialty flasks, transfer caps and the Rapid Clear Cap for media clarification.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.