Learn More
Biotium Biotin-4-Fluorescein
Supplier: Biotium 90062
Biotin-4-fluorescein (N-(5-aminopentyl)biotinamide, trifluoroacetic acid salt) has been reported to be a superior fluorescent biotin derivative for accurate measurement of avidin and streptavidin in crude biological samples. Binding of biotin to avidin and streptavidin is high affinity, fast and noncooperative. Properties: Ex/ Em = 494/523 nm; Yellow solid soluble in DMSO, or pH >7 buffers; Store at 4°C and protect from light; C33H32N4O8S; MW: 644.7
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.