Learn More
Biotium Biotin Picolyl Azide
Supplier:  Biotium 92186
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Biotin picolyl azide reacts with alkyne to form 1,2,3-triazole by 1,3-dipolar Huisgen cycloaddition through the use of a much lower copper (I) concentration without sacrificing reaction efficiency.
Catalog No. 50-196-4916