Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-carboxyethyl) Isocyanurate 98.0+%, TCI America™
SDP

Catalog No. p-7131500 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 2904-40-7
Molecular Formula C9H11N3O7
Molecular Weight (g/mol) 273.201
MDL Number MFCD04038422
InChI Key JCUMQTAAEUDUPK-UHFFFAOYSA-N
Synonym Isocyanuric Acid Bis(2-carboxyethyl) Ester
PubChem CID 223763
IUPAC Name 3-[3-(2-carboxyethyl)-2,4,6-trioxo-1,3,5-triazinan-1-yl]propanoic acid
SMILES C(CN1C(=O)NC(=O)N(C1=O)CCC(=O)O)C(=O)O

Specifications

Melting Point 289°C
Color White
Quantity 25 g
Formula Weight 273.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-carboxyethyl) Isocyanurate
EINECSNumber (5)-1063
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.