Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-formylphenyl) Ether 98.0+%, TCI America™
SDP

Catalog No. p-7119180 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
10 g

Chemical Identifiers

CAS 49590-51-4
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD00191616
InChI Key LMJZLKFWMQOYKU-UHFFFAOYSA-N
PubChem CID 614688
IUPAC Name 2-(2-formylphenoxy)benzaldehyde
SMILES C1=CC=C(C(=C1)C=O)OC2=CC=CC=C2C=O

Specifications

Melting Point 78°C
Color White-Yellow
Boiling Point 182°C
Quantity 1 g
Synonym 2,2′C-Diformyldiphenyl Ether, 2,2′C-Oxydibenzaldehyde
Formula Weight 226.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-formylphenyl) Ether
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.