Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-iodophenyl)amine 98.0+%, TCI America™
SDP

Catalog No. p-7119715 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 20255-70-3
Molecular Formula C12H9I2N
Molecular Weight (g/mol) 421.02
InChI Key SJLIWXXAQHNDHM-UHFFFAOYSA-N
Synonym 4,4′C-Diiododiphenylamine
PubChem CID 12950430
IUPAC Name 4-iodo-N-(4-iodophenyl)aniline
SMILES C1=CC(=CC=C1NC2=CC=C(C=C2)I)I

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 1 g
Formula Weight 421.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-iodophenyl)amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.