Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(pentafluorophenyl)phenylphosphine 98.0+%, TCI America™
SDP

Catalog No. p-7123617 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 5074-71-5
Molecular Formula C18H5F10P
Molecular Weight (g/mol) 442.20
MDL Number MFCD00000291
InChI Key OYNXPGGNQMSMTR-UHFFFAOYSA-N
Synonym bis pentafluorophenyl phenylphosphine, dftpp, decafluorotriphenylphosphine, bis perfluorophenyl phenyl phosphine, bis 2,3,4,5,6-pentafluorophenyl-phenylphosphane, phosphine,bis 2,3,4,5,6-pentafluorophenyl phenyl, bis 2,3,4,5,6-pentafluorophenyl phenyl phosphane, ultramark 443, bis pentafluorophenyl phenyl phosphite, bis pentafluorophenyl phenyl phosphine
PubChem CID 78757
IUPAC Name bis(2,3,4,5,6-pentafluorophenyl)(phenyl)phosphane
SMILES FC1=C(F)C(F)=C(P(C2=CC=CC=C2)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 105°C
Quantity 200 mg
Formula Weight 442.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(pentafluorophenyl)phenylphosphine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.