Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(tetrabutylammonium) Dihydrogen Pyrophosphate 96.0+%, TCI America™
SDP

Catalog No. p-7124076 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 857447-79-1
Molecular Formula C32H74N2O7P2
Molecular Weight (g/mol) 660.899
InChI Key WPGBCELIUCBRKN-UHFFFAOYSA-L
Synonym Bis(tetrabutylammonium) Dihydrogen Diphosphate
PubChem CID 11342793
IUPAC Name [hydroxy(oxido)phosphoryl] hydrogen phosphate;tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.OP(=O)([O-])OP(=O)(O)[O-]

Specifications

Color White
Quantity 200 mg
Formula Weight 660.90
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(tetrabutylammonium) Dihydrogen Pyrophosphate
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.