Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bis(trifluoromethanesulfonyl)imide 99.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Melting Point | 56°C |
Color | White |
Boiling Point | 92°C |
Quantity | 5 g |
UN Number | 3261 |
Formula Weight | 281.14 |
Percent Purity | ≥99.0% (T) |
Physical Form | Crystalline Powder |
Chemical Name or Material | Bis(trifluoromethanesulfonyl)imide |
Safety and Handling
TSCA | Yes |
Recommended Storage | Refrigerator |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction