Learn More
AdipoGen Bisindolylmaleimide I (5 mg)
SDP
Supplier:  AdipoGen AGCR10009M005
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS: 133052-90-1. Formula: C25H24N4O2. MW: 412.5. Cell permeable kinase inhibitor with improved selectivity for protein kinase C (PKC) (Ki = 10 nM). Competetive inhibitor for the ATP-binding site of PKC. Anti-inflammatory. Binds to P-glycoprotein. Telomerase activity inhibitor. Potent glycogen synthase kinase-3 (GSK-3) inhibitor. Necrosis inhibitor. Blocks hERG potassium channels. Promotes osteoblastogenesis in human mesenchymal stem cells.
Catalog No. 50-205-3757