Learn More
AdipoGen Bisindolylmaleimide I HCl
SDP
Supplier:  AdipoGen AGCR10110M005
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 176504-36-2. Formula C25H24N4O2 . HCl. MW 412.5 . 36.5. Cell permeable kinase inhibitor with improved selectivity for protein kinase C PKC Ki = 10 nM. Competitive inhibitor for the ATP-binding site of PKC. Anti-inflammatory. Binds to P-glycoprotein. Telomerase activity inhibitor. Potent glycogen synthase kinase-3 GSK-3 inhibitor. Necrosis inhibitor. Blocks hERG potassium channels. Promotes osteoblastogenesis in human mesenchymal stem cells.
Catalog No. 50-256-2365