Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Bismuth Nitrate
SCI_ED

Catalog No. S25198B Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
Packaging:

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS

10035-06-0

Molecular Formula

BiH10N3O14

Molecular Weight (g/mol)

485.07

MDL Number

MFCD00149157

InChI Key

FBXVOTBTGXARNA-UHFFFAOYSA-N

Synonym

bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate

PubChem CID

160911

IUPAC Name

bismuth(3+) pentahydrate trinitrate

SMILES

O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color

White

Physical Form

Crystalline

Quantity

100 g

Assay Percent Range

98% w/v

Linear Formula

Bi(NO3)3·5H2O

Formula Weight

485.11

Grade

Laboratory

Packaging

Lab Grade - Crystals

Density

2.83g/cmsuperscript_3;

Chemical Name or Material

Bismuth Nitrate

ShelfLife

One year

DOTInformation

DOT Class 5.1, II, : Oxidizer

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.