Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bismuth titanium isopropoxide, 5% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
Click to view available options
Quantity:
25 mL
100 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Packaged under Argon
Specifications
Specifications
Color | Yellow |
Packaging | ChemSeal™ bottles |
Linear Formula | BiTi[OCH(CH3)2]7 |
Quantity | 25 mL |
UN Number | UN1219 |
Sensitivity | Moisture Sensitive |
Formula Weight | 670.48 |
Physical Form | Liquid |
Chemical Name or Material | Bismuth titanium isopropoxide |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement H225-H319-H336 Highly flammable liquid and vapor. Causes serious eye irritation. May cause drowsiness or dizziness. |
Precautionary Statement | GHS P Statement P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL |
storageNote1 |
|
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction