Learn More
Agilent Technologies BL21-GOLD(DE3)PLYSS C.CELLS
Supplier:  Agilent Technologies 230134
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
BL21-Gold (DE3)pLysS Competent Cells Provide high-level protein expression and Allow direct cloning of many expression constructs. Hte phenotype increases transformation efficiency to 1 x 108 of supercoiled DNA. Used for toxic proteins.
Catalog No. 50-125-349
missing translation for 'QuantityLeft'