Learn More
Aqua Phoenix™ Blast Off! Deluxe Kit Components
Shop All AquaPhoenix Scientific Products

Specifications
Product Type
White vinegar
Grade
K to 5
For Use With (Application)
Education Kit
Capacity (English)
1 gal.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.