Learn More
Gold Biotechnology Inc Blasticidin S HCl 100 mg
Supplier: Gold Biotechnology Inc B800100
Blasticidin S is an aminoacylnucleoside antibiotic isolated from Streptomyces griseochromogenes. It shows high efficacy against both eukaryotic and prokaryotic cells. Aside from its antimicrobial properties, blasticidin has displayed antiviral, antitumor, and antifungal properties and has been used as a selection agent. It has been used to select for cells containing the bls, bsr, or BSD resistance gene as well as HEK293-T cells and HEK-D5 cells. Additionally, blasticidin is effective in preventing cell culture contam
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.