Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Blebbistatin, Tocris Bioscience™
GSA_VA

Catalog No. 176010R Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective inhibitor of myosin II ATPase activity

(±)-Blebbistatin is a selective inhibitor of myosin II ATPase activity; inhibits contraction of the cleavage furrow without disrupting mitosis or contractile ring assembly. (?)-Blebbistatin rapidly and reversibly blocks cell blebbing, and disrupts directed cell migration and cytokinesis in vertebrate cells. (?)-Blebbistatin inhibits arterial thrombosis development, reduces infiltration of inflammatory cells, and reduces damage to vascular tissue in vivo in a mice model of arterial thrombosis.

Active and Inactive Enantiomer also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 674289-55-5
Molecular Formula C18H16N2O2
Molecular Weight (g/mol) 292.338
InChI Key LZAXPYOBKSJSEX-UHFFFAOYSA-N
Synonym +/--blebbistatin, blebbistatin, 3a-hydroxy-6-methyl-1-phenyl-2,3-dihydropyrrolo 2,3-b quinolin-4-one, 3a-hydroxy-6-methyl-1-phenyl-1,2,3,3a-tetrahydro-4h-pyrrolo 2,3-b quinolin-4-one, +--blebbistatin, 3a-hydroxy-6-methyl-1-phenyl-2h,3h-pyrrolo 2,3-b quinolin-4-one, 1-phenyl-1,2,3,4-tetrahydro-4-hydroxypyrrolo 2.3-b-7-methylquinolin-4-one, inverted exclamation marka-blebbistatin, +/-bebbistatin, --blebbistcitin
PubChem CID 3476986
ChEBI CHEBI:75379
IUPAC Name 3a-hydroxy-6-methyl-1-phenyl-2,3-dihydropyrrolo[2,3-b]quinolin-4-one
SMILES CC1=CC2=C(C=C1)N=C3C(C2=O)(CCN3C4=CC=CC=C4)O

Specifications

Quantity 10 mg
Formula Weight 292.34
Percent Purity >98%
Chemical Name or Material (±)-Blebbistatin
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.