Learn More
RIGAKU REAGENTS Block, Wizard PEG Ion Bundle, 192 formulations, 1.7 mL volumes
Supplier:  RIGAKU REAGENTS 1010286
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
The PEG Ion series matrix combines the crystallizing power of polyethylene glycols (PEGs) with salts that are commonly found in crystallization conditions. Use PEG Ion during the coarse screening phase of biological macromolecules (proteins, nucleic acids, peptides, and combinations thereof).
Catalog No. 50-149-2092