Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMeS-p-A 95.0+%, TCI America™
SDP

Catalog No. p-7124072 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 1678513-97-7
Molecular Formula C8H12N2O4S2
Synonym 1,4-Diamino-2,5-bis(methylsulfonyl)benzene, 2,5-Bis(methylsulfonyl)-1,4-phenylenediamine

Specifications

Quantity 200 mg
Formula Weight 264.31
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material BMeS-p-A
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.