Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMS 961, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective RARγ agonist (EC50 values are 30 and 1000 nM at RARγ and RARβ respectively). Displays no activity at RARα receptors.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 399.46 |
Percent Purity | >97% |
Chemical Name or Material | BMS 961 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction