Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMS 986187, >98%, Tocris Bioscience™

Catalog No. p-7121250 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent δ-opioid receptor positive allosteric modulator

Potent δ-opioid receptor positive allosteric modulator (EC50 = 30 nM). Exhibits 100-fold selectivity for δ over μ-opioid receptor. Enhances affinity of [Leu5]-Enkephalin (Cat.No. 1889), SNC 80 (Cat.No. 0764) and TAN 67 (Cat.No. 0921) for the δ-opioid receptor.

Chemical Identifiers

CAS 684238-37-7
Molecular Formula C31H34O4
Molecular Weight (g/mol) 470.609
InChI Key UEKIYVKPQNKSDI-UHFFFAOYSA-N
Synonym 3,3,6,6-tetramethyl-9-4-2-methylbenzyl oxy phenyl-3,4,5,6,7,9-hexahydro-1h-xanthene-1,8 2h-dione, 3,3,6,6-tetramethyl-9-4-2-methylbenzyloxy phenyl-3,4,5,6-tetrahydro-9h-xanthene-1 2h ,8 7h-dione, 3,3,6,6-tetramethyl-9-4-2-methylphenyl methoxy phenyl-4,5,7,9-tetrahydro-2h-xanthene-1,8-dione, 3,4,5,6,7,9-hexahydro-3,3,6,6-tetramethyl-9-4-2-methylphenyl methoxy phenyl-1h-xanthene-1,8 2h-dione
PubChem CID 17379334
IUPAC Name 3,3,6,6-tetramethyl-9-[4-[(2-methylphenyl)methoxy]phenyl]-4,5,7,9-tetrahydro-2H-xanthene-1,8-dione
SMILES CC1=CC=CC=C1COC2=CC=C(C=C2)C3C4=C(CC(CC4=O)(C)C)OC5=C3C(=O)CC(C5)(C)C

Specifications

Linear Formula C31H34O4
Quantity 10 mg
Solubility Information Soluble to 20mM in DMSO
Formula Weight 470.61
Chemical Name or Material 3,4,5,6,7,9-Hexahydro-3,3,6,6-tetramethyl-9-[4-[(2-methylphenyl)methoxy]phenyl]-1H-xanthene-1,8(2H)-
Grade >98%
Recommended Storage Store at 4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.