Learn More
Medchemexpress LLC HY-101908 1mg Medchemexpress, BMS CCR2 22 CAS:445479-97-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1019081MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-101908 1mg BMS CCR2 22 CAS:445479-97-0 BMS CCR2 22 is a potent, specific and high affinity CC-type chemokine receptor 2 (CCR2) antagonist with excellent binding affinity (binding IC50 of 5.1 nM) and potent functional antagonism (calcium flux IC50 of 18 nM and chemotaxis IC50 of 1 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7911
missing translation for 'QuantityLeft'