Learn More
Echelon Biosciences Research Labs BODIPY FL-PI(5)P 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs C05F6100UG
Encompass
BODIPY FL PI(5)P (BODIPY FL Phosphatidylinositol 5-phosphate) is a water soluble analog of PI(5)P labeled with the green fluorophore, Bodipy-FL at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 5-phosphate (PI(5)P) is a rare lipid found in the nucleus that can bind the PHD finger of the nuclear adapter protein ING2. PI(5)P is phosphorylated to PI(4,5)P2 by Type II PIP kinases. This item is non returnable
Catalog No. 50-720-073
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.