Learn More
Electron Microscopy Sciences Bone Shears Stainless Steel, 5 1/2" OAL
Supplier: Electron Microscopy Sciences 7289405
Made from stainless steel with a removable bar spring; perfect for cutting cartilage and bone.
OAL: 5 ½"
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.