Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Boric Acid
SCI_ED

Catalog No. p-4416655 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade
Reagent Grade

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Color White
Density ∽1.44g/mL
Packaging Lab Grade
Assay Percent Range >95% w/v
Quantity 100 g
Physical Form Solid Crystal/Powder
Linear Formula H3BO3
Formula Weight 61.83
Chemical Name or Material Boric Acid
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.