Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Boric Acid, MP Biomedicals™
Shop All MP Biomedicals, Inc Products
Click to view available options
:
Quantity: 100 g
Description
- Boric Acid is used for weatherproofing wood and fireproofing fabrics.
- It is used as a preservative.
- It is also used in the manufacturing of cements, crockery, porcelain, enamels, glass, borates, leather, carpets, hats, soaps, and artificial gems.
- Boric Acid is used in nickeling baths, cosmetics; printing and dyeing, painting; photography; for impregnating wicks; electric condensers; hardening steel.
- It is also used as insecticide for cockroaches and black carpet beetles.
- In the research labs it is used as an astringent, antiseptic, antibacterial and antifungal agent.
- Used in buffers and for calibration of polyacrylamide gel columns for the separation of oligonucleotides by capillary electrophoresis.
- The Vapor Pressure of Boric Acid is 1.6X10-6 mm Hg (25°C) (2.136X10-4 Pa); log P (in Pa) = 26.83 - 9094/T where T is °K
Specifications
Specifications
pH | 3.8 to 4.8 (3.3% aq. soln.) |
Melting Point | 340°F |
CAS | 10043-35-3 |
Boiling Point | 572°F at 760 mm Hg (Decomposition) |
Density | 1.435 g/mL at 68°F |
Vapor Pressure | 1.6X10-6 mm Hg at 25°C |
Quantity | 100 g |
Physical Form | Powder |
Synonym | Boracic Acid, Orthboric Acid |
Solubility Information | 10 to 50 mg/mL at 66°F |
Show More |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction