Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid Solution with Indicator, Ricca Chemical
SDP

Catalog No. p-20120 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
1 L
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle

Chemical Identifiers

CAS 10043-35-3, 7732-18-5, 64-17-5, 67-56-1, 67-63-0, 63451-28-5
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

CAS Min % 3.76, 94.0, 0.2, 0.01, 0.1, 1.0E-4
CAS Max % 3.92, 98.0, 0.3, 0.02, 0.2, 1.0E-4
Packaging Natural Poly Bottle
Quantity 1 L
Chemical Name or Material Boric Acid Solution with Indicator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.