Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Boron Trifluoride - Ethyl Ether Complex 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 mL
100 mL
500 mL
Specifications
Specifications
Melting Point | -60°C |
Color | Yellow |
Boiling Point | 124°C |
Quantity | 25 mL |
UN Number | 2604 |
Formula Weight | 141.93 |
Percent Purity | ≥98.0% (W) |
Physical Form | Liquid |
Chemical Name or Material | Boron Trifluoride - Ethyl Ether Complex |
Safety and Handling
EINECSNumber | (2)-2107 |
RTECSNumber | ED2275000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction