Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Borosilicate Glass Test Tubes with Rim
SDPSCI_ED

Safely packaged test tubes with good clarity

Supplier:  United Scientific™ TT9800M

 View more versions of this product

Catalog No. S01069


missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy

Description

Description

  • Pre-cleaned borosilicate glass test tubes with rims
  • Tubes can be used for hot or cold applications, are reusable, and can be autoclaved
Specifications

Specifications

16 mL
Borosilicate Glass
72/Pk.
Autoclavable
16 mm
Test Tube
125 mm
Cylindrical
Non-sterile
No
Not Included
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.