Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Roylco™ Botanical Paper Cut-outs

Click to view available options
:
Botanical Paper Cut-outs; No. of Pages: 210 Pages
Description
- Wide range of leaf and flower shapes
- Ideal for making collages, decorating posters, or making 3D flowers
- Made of card stock
- Double sided, two tone colors
- Acid-free
Specifications
Specifications
Grade | 1+ |
Format | Kit |
Includes | 96 Leaves, 114 Flowers, Idea Guide |
No. of Pages | 210 Pages |
Languages | English, Spanish, French, German |
Subject | Plants |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction