Learn More
Hach Company Bottle, BOD, Black, 300 mL
Supplier: Hach Company 2086047
The standard for BOD analysis. Borosilicate glass bottle with flared lip and ground-glass stopper to prevent air entrapment and for water seal. Frosted marking area. Stopper and an opaque black cap included (60 and 300 mL sizes). Serialized bottles have silk-screened, sequential numbers and include a bar code.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.