Learn More
Wilmad Labglass BOTTLE SPECIFIC GRAV 25ML ADJ
Supplier:  Wilmad Labglass LG3540106
Encompass_Preferred
Bottle, Specific Gravity, Gay-Lussac, Adjusted. Capacity: 25mL; Joint Size: [ST] 10/18. Adjusted at 20degreeC. The capillary stoppers are not interchangeable since the final adjustment for volume is made on the stopper.
Catalog No. 50-805-719
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.