Learn More
Corning™ Bottle Top Dispenser (BTD) PTFE Plunger
PTFE plunger for BTD and dispensing reagents
$50.90 - $125.80
Specifications
For Use With (Application) | Dispensing Organic and Inorganic Liquids |
---|---|
Includes | Four adapters |
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
10-320-162
|
Corning™
6861 |
2.5 mL |
Case for $50.90
|
|
|||||
10-320-163
|
Corning™
6862 |
5 mL |
Case for $54.70
|
|
|||||
10-320-164
|
Corning™
6863 |
10 mL |
Case for $63.70
|
|
|||||
10-320-165
|
Corning™
6864 |
25 mL |
Case for $82.60
|
|
|||||
10-320-166
|
Corning™
6865 |
50 mL |
Case for $100.60
|
|
|||||
10-320-167
|
Corning™
6866 |
100 mL |
Case for $125.80
|
|
|||||
Description
- Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
- Comes with four adapters with different diameters for universal use with multiple bottle sizes
- Telescopic suction tube can be adjusted smoothly to different bottle heights
- Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
- Every dispenser is individually tested and delivered with a certificate of performance
- For dispensing organic and inorganic liquids
Specifications
Dispensing Organic and Inorganic Liquids | |
Four adapters |