Learn More
Rockland Immunochemicals BOVINE/CALF BLOOD in Sodium Citrate - R200-0100
Supplier: Rockland Immunochemicals R2000100
Citrated bovine blood is aseptically collected and prepared to order from our colony of animals. These cell suspensions are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Bovine red blood cells are perishable and are collected and processed upon receipt of your order.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.