Learn More
Creative Biomart Pegademase, Bovine adenosine deaminase
Supplier:  Creative Biomart THP0104
Encompass_Preferred
Bovine adenosine deaminase derived from bovine intestine that has been extensively pegylated for extended serum half life.
Catalog No. 50-208-4155
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.