Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bovine Albumin Cohn

Catalog No. MP08810012 Shop All MP Biomedicals, Inc Products
Click to view available options
:

Heat shocked, precipitated by cold ethanol and crystallized.

  • Used to solubilize lipids, and used as blocking agent in Western blots or ELISA applications

Specifications

Concentration

98% protein

Storage Requirements

Store at 2° to 8°C

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.