Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bovine Albumin Cohn Modified, Lyophilized

Catalog No. p-7112912 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
50g
100g

Used to solubilize lipids, as blocking agent in Western blots or ELISA applications.

  • Cohn modified, heat-shocked fractionated

Specifications

Concentration Protein: >96%
For Use With (Application) Immunoassay, Mammalian Cell Culture
Format Powder
pH Range 6.84
Purity >96%By Protein Electrophoresis (Albumin Fraction V)
Quantity 50g
Source Bovine
Storage Requirements Store at 2° to 8°C
Product Type Bovine albumin
Protein Subtype Albumins

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.