Learn More
Kingfisher Biotech Inc Bovine CCL8 - 500ug
SDP
Supplier:  Kingfisher Biotech Inc RP1903B500
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
The Bovine CCL8 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine CCL8 applications are for cell culture, ELISA standard, and Western Blot Control. Bovine CCL8 yeast-derived recombinant protein can be purchased in multiple sizes. Bovine CCL8 Specifications: (Molecular Weight: 8.6 kDa) (Amino Acid Sequence: QPDSVSTPITCCFSVINGKIPFKKLDSYTRITNSQCPQEAVIFKTKADRDVCADPKQKWVQTSIRLLDQKSRTPKP (76)) (Gene ID: 281044). For research use only. Made in the USA
Catalog No. 50-253-2606
missing translation for 'QuantityLeft'