Learn More
Kingfisher Biotech Inc Bovine IL-36 alpha - 5ug
SDP
Supplier:  Kingfisher Biotech Inc RP1933B005
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
The Bovine IL-36 alpha yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine IL-36 alpha applications are for cell culture, ELISA standard, and Western Blot Control. Bovine IL-36 alpha yeast-derived recombinant protein can be purchased in multiple sizes. Bovine IL-36 alpha Specifications: (Molecular Weight: 16.4 kDa) (Amino Acid Sequence: NIQDINHLVW VLQGQALTAV PRKDQMDPVT VTLVSCKYTE TLEKGRGNPV YLGLKEPELC LFCTKVKGQP TLQLQERNIM DLYHQTEPVK PFLFYHDQNG RASSFESVAF PGWFIGSCSC GGCPVIITQE LGKIYTTDFG FTVLQP (146)) (Gene ID: 523429). For research use only. Made in the USA
Catalog No. 50-253-2597
missing translation for 'QuantityLeft'