Learn More
Kingfisher Biotech Inc Bovine Osteopontin - 500ug
SDP
Supplier:  Kingfisher Biotech Inc RP2089B500
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
The Bovine Osteopontin yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine Osteopontin applications are for cell culture, ELISA standard, and Western Blot Control. Bovine Osteopontin yeast-derived recombinant protein can be purchased in multiple sizes. Bovine Osteopontin Specifications: (Molecular Weight: 29.2 kDa) (Amino Acid Sequence: LPVKPTSSGS SEEKQLNNKY PDAVATWLKP DPSQKQTFLA PQNSVSSEET DDNKQNTLPS KSNESPEQTD DLDDDDDNSQ DVNSNDSDDA ETTDDPDHSD ESHHSDESDE VDFPTDIPTI AVFTPFIPTE SANDGRGDSV AYGLKSRSKK FRRSNVQSPD ATEEDFTSHI ESEEMHDAPK KTSQLTDHSK ETNSSELSKE LTPKAKDKNK HSNLIESQEN SKLSQEFHSL EDKLDLDHKS EEDKHLKIRI SHELDSASSE VN (262)) (Gene ID: 281499). For research use only. Made in the USA
Catalog No. 50-253-2455
missing translation for 'QuantityLeft'