Learn More
3M™ Bovine Total Milk Protein ELISA Kit, E96MLK, Sold by AquaPhoenix Scientific
Detects bovine total milk proteins in clean-in-place water (CIP) final rinse water, environmental swab samples, food ingredients and processed food products
Supplier: 3M™ E96MLK
Description
- Designed to detect processed and unprocessed target allergen proteins
- Test can accommodate environmental swab, CIP rinse water and food samples
- Universal extraction protocol streamlines testing for multiple analytes, saving reagents and time
Specifications
Bovine total milk protein | |
ELISA kit | |
96-well plate | |
50 min. |