Learn More
Tecan Protein Fragment BoxA from HMGB1, human & mouse, LPS-free
Supplier: Tecan REHM012
BoxA from HMGB1, human & mouse, LPS-free. 100 ug. BoxA is one of the highly conserved DNA binding domains of HMGB1 protein (HMG boxes). It is an antagonist for HMGB1 binding to its receptor RAGE, and inhibits HMGB1 cytokine effects in vivo. The BoxA we provide has no difference from the sequence of the human or mouse HMGB1, and as a self-antigen elicits very low immunological responses. BoxA protects against sepsis in a mouse peritonitis model, and from hepatitis in a mouse model of HBV infection. Dendritic cells promote their own maturation and T cell proliferation, survival and Th1 differentiation by secreting HMGB1, and BoxA inhibits these activities. BoxA also inhibits the chemotactic activity of HMGB1 on endothelial cells and fibroblasts, and the mitogenic activity of HMGB1 for stem cells. Distributed by IBL. Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.