Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Caution: Hazardous Waste Storage Area… Signs

Catalog No. p-167737 Shop All Brady Corporation Products
Click to view available options
Material:
Height (English):
Width (English):

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents.

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads “CAUTION HAZARDOUS WASTE STORAGE AREA UNAUTHORIZED PERSONS KEEP OUT”
  • Black on yellow
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene
  • B-120
    • Premium fiberglass — fiberglass reinforced polyester
  • B-302
    • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic

    Specifications

    Certifications/Compliance

    ANSI Z535, OSHA 1910.145, ISO 9000, ASTM D 1593, ASTM D 638, ASTM D 638, ASTM D 646

    Material

    Fiberglass

    Quantity

    1 Ea.

    Tensile Strength

    9000 psi

    Description

    Premium Fiberglass

    Mounting Type

    Surface Mount

    For Use With (Application)

    Industrial, utility, commercial and institutional environment

    Height (English)

    10 in.

    Height (Metric)

    25.4 cm

    Max. Temperature (English)

    193°F

    Min. Temperature (English)

    -40°F

    Thickness

    0.1 in.

    Width (English)

    14 in.

    Width (Metric)

    35.5 cm

    Temperature Range (Metric)

    -40°C to 90°C

    Temperature Range (English)

    -40°F to 193°F

    Displayed Text

    CAUTION Hazardous Waste Storage Area Unauthorized Persons Keep Out

    Product Type

    Chemical and Hazardous Materials Sign

    Color

    Black on Yellow

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.