Learn More
Brady™ Markware™ Software Kit
Catalog No. 19000065 Shop All Brady Corporation Products
Click to view available options
:
Markware™ Software Kit provides all safety and facility identification needs in one package.
  • Use MarkWare™ Software on your PC to design professional-looking signs, labels and tags
  • The label-making template wizard walks you through the creation process step-by-step, making the program easy to use without a lot of training
  • Print directly to a labeling system from Brady™ or most Windows™ inkjet/laser printers
  • Use preformatted templates provided for quick and easy label creation or freely customize label layouts to your own specifications
Specifications
Product Type
Markware Software Kit
For Use With (Equipment)
BBP31 Sign and Label Printer, BBP33 Label Printer, BMP71 Label Printer, GlobalMark Printer, HandiMark Printer, MiniMark Printer, PowerMark Sign and Label Printer
Sterility
Non-sterile
Certifications/Compliance
ANSI;NFPA Compliant;OSHA
For Use With (Application)
Energy Source Labeling, Facility, Safety and Maintenance Indentificaiton, Hazardous Material Labeling, NFPA Compliant, Production/ Bar Code Labeling Software, Rating Plate Identification, Right to Know Labeling, Visual Equipment Controls, Warehouse Bin and Shelf Labeling
Quantity
1 Ea.